Learn More
Fisherbrand™ Polypropylene Universal Pipette Tips
Protect your samples, pipettes and results.by using Fisherbrand™ Polypropylene Universal Pipette Tips. These versatile tips will prevent cross-contamination.
45.00€ - 133.56€
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||
11867292
|
Fisherbrand™
|
5 to 200 μL | Natural |
133.56€
Pack of 700 |
In Stock
Log in to see stock. |
||||
11851843
|
Fisherbrand™
|
5 to 200 μL | Lace White |
54.48€
Pack of 1000 |
In Stock
Log in to see stock. |
||||
10380691
|
Fisherbrand™
|
300 μL | Natural |
46.06€
Pack of 1000 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||||
11857272
|
Fisherbrand™
|
10 mL | Natural |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
11887292
|
Fisherbrand™
|
1 to 5 mL | Natural |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
- Polypropylene tip
- Superior accuracy and precision
- Autoclavlable
- Variety of volume ranges
Specifications
Sterile | |
Polypropylene |