Learn More
Buerkle™ Polypropylene Universal Graduated Beakers
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
400 mL
1000 mL
Unit Size:
Each
Description
- Can be used as disposable
- Manufactured from transparent polypropylene
- Autoclavable
- Space-saving: beakers stack in one another
Specifications
Specifications
| Color | Transparent |
| Diameter (Metric) Base | 72 mm |
| Diameter (Metric)Top | 113 mm |
| Capacity (Metric) | 1000 mL |
| Material | Polypropylene |
| Increments | 50 mL |
| Quantity | Each |
| Type | Beaker |
| Autoclavable | Autoclavable |
| Disposable | Single-use |
| Show More |