Learn More
Buerkle™ Polypropylene Universal Graduated Beakers
Product Code. 10066631 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
400 mL
1000 mL
Unit Size:
Each
Attention! Make sure to pay attention to the List of Chemical Resistance!
Product Code. 10066631
Brand: Buerkle™ 71090400
Only null left
Add to basket
Add to basket
Attention! Make sure to pay attention to the List of Chemical Resistance!
Polypropylene Universal Graduated Beakers with indestructable graduation and three specially formed spouts for easy pouring of liquids.
  • Can be used as disposable
  • Manufactured from transparent polypropylene
  • Autoclavable
  • Space-saving: beakers stack in one another
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Diameter (Metric) Base 51 mm
Diameter (Metric)Top 83 mm
Capacity (Metric) 400 mL
Material Polypropylene
Increments 20 mL
Quantity Each
Type Beaker
Autoclavable Autoclavable
Disposable Single-use
Graduated Yes