Learn More
Semadeni™ Polypropylene Tweezer
Product Code. 11707202
Click to view available options
Length (Metric):
130 mm
Unit Size:
Each
Product Code. 11707202
Brand: Semadeni™ 1648
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Order Info
dimLengthMetric
Specifications
Tip Style Stump
Material Polypropylene
Length (Metric) 130 mm
Product Type Tweezers
Quantity (No. per Case) 100/Pack