Learn More
Camlab™ Polypropylene Tube Rack
Product Code. p-8000124
Encompass
Click to view available options
Color:
Blue
Red
Unit Size:
Each
Product Code. 11787586
Brand: Camlab™ RTP711096B
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Product Type Tube Rack
Color Blue
Holds Microtubes (0.5 and 2.0 mL)
Material Polypropylene
Positions 96 and 48
For Use With (Equipment) For both PCR and microcentrifuge tubes
Quantity 1 Each
Width (Metric) 68 mm
Height (Metric) 21 mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.