Learn More
Kartell™ Polypropylene Micropipette Tips with Filter
Designed with hydrophobic PE filters that act as a barrier against aerosol contamination, preventing cross-contamination of samples and micropipettes. Kartell™ Polypropylene Micropipette Tips with Filter are sized to completely empty the tip in the event of over-aspiration.
Brand:  Kartell™ 0963300
Additional Details : Weight : 0.09000kg
Product Code. 10276571
  • 225.00€ / Pack of 960
Estimated Shipment: 07-06-2024
Description
  • DNA-free, RNA-free, cadmium-free, apyrogenic
  • Polypropylene construction; includes a filter
  • Sterilized by gamma radiation; Autoclavable at 121°C for 20 minutes
  • Use in molecular biology, cell cultures, bacteriology, virology, immunology and radioimmunology