Learn More
Poulten Graf™ Polypropylene Threaded Adapter
Click to view available options
Thread Size:
GL 32, 38 mm
GL 32, 40 mm
GL 32, 52 mm
Unit Size:
Each
Description
- Accessory
Specifications
Specifications
| Product Type | Thread Adapter |
| Material | Polypropylene |
| For Use With (Equipment) | Optifix Dispensers, Adapters Distributor Collar, GL32 Screw |
| Quantity | 1 Ea. |
| Thread Size | GL 32, 38 mm |