Learn More
Fisherbrand™ Super Slim Extra Fine Pipette Tips
Use Fisherbrand™ Super Slim Extra Fine Pipette Tips in the laboratory since they are the critical link in ensuring that the correct volume of liquid is aspirated and dispensed and that the liquid is not contaminated in the process.
Brand:  Fisherbrand™
Additional Details : Weight : 0.72000kg
 View more versions of this product
Product Code. 11807402
  • 65.24€ / Pack of 1000
In Stock
Description
  • Autoclavable
  • Super slim
  • Polypropylene
Specifications
Specifications
5 to 500 μL
Translucent
117mm
Finely Tapered
Polypropylene
Sterile
Autoclavable
Individually Wrapped
Super Slim