Learn More
Fisherbrand™ Super Slim Extra Fine Pipette Tips
Description
- Autoclavable
- Super slim
- Polypropylene
Specifications
Specifications
| Volume (Metric) | 200 to 2500 μL |
| Sterility | Non-sterile |
| Color | Translucent |
| Autoclavable | Autoclavable |
| Length (Metric) | 95mm |
| Tip Style | Finely Tapered |
| Product Line | Super Slim |
| Material | Polypropylene |