Learn More
Fisherbrand™ Super Slim Extra Fine Pipette Tips
Use Fisherbrand™ Super Slim Extra Fine Pipette Tips in the laboratory since they are the critical link in ensuring that the correct volume of liquid is aspirated and dispensed and that the liquid is not contaminated in the process.
Brand: Fisherbrand™
Additional Details : Weight : 0.72000kg
Description
- Autoclavable
- Super slim
- Polypropylene
Specifications
5 to 500 μL | |
Translucent | |
117mm | |
Finely Tapered | |
Polypropylene |
Sterile | |
Autoclavable | |
Individually Wrapped | |
Super Slim |