Learn More
Multiroir™ Polypropylene Storage Boxes
Product Code. 11872702
Click to view available options
Capacity (Metric):
25 L
30 L
Unit Size:
Each
Product Code. 11872702
Brand: Multiroir™ 18476
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Store and transport items securely in high-capacity Multiroir™ Polypropylene Storage Boxes. The boxes have snap-on, tight-fitting lids and have no handles.
  • Translucent Polypropylene
  • Large capacity
  • Without handles
Specifications
Capacity (Metric) 25 L
Color Translucent
Material Polypropylene
Shape Rectangular
Product Type Storage Box
Quantity 1 Each
Dimensions (L x W x H) 550 x 390 x 160 mm
Length (Metric) 550 mm
Width (Metric) 390 mm
Width (Metric) Interior 390 mm
Height (Metric) 160 mm
Weight (Metric) 0.938 kg