Learn More
Witeg™ Polypropylene Screw Caps
Click to view available options
Thread Size (Metric):
GL 25
GL 45
GL 80
GL100
Color:
Neutral
Red
Yellow
Unit Size:
Each
Pack of 10
Description
- Autoclavable up to 140°C
- GL 45 Thread
Specifications
Specifications
| Coating | PTFE |
| Color | Red |
| Includes | PTFE coated seal |
| Material | Polypropylene |
| No. per Pack | 10Pack |
| Quantity | 10Pack |
| Screw Color | Red |
| Screw Material | Polypropylene |
| Thread Size (Metric) | GL 45 |
| Liner Material | PTFE |
| Show More |