Learn More
Cole-Parmer™ Polypropylene Screen Disc
Product Code. 11777113
Change view
Click to view available options
Pore Size:
149 μm
500 μm
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
11777113 500 μm Pack of 10
11797113 149 μm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11777113 Supplier Cole-Parmer™ Supplier No. 0663563
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Material Polypropylene
Pore Size 500 μm
Quantity 10Pack
Thickness (Metric) 610 μm
Type Screen Disc
Diameter (Metric) 55 mm