Learn More
Vitlab™ Polypropylene Scoop
Use these corrosion resistant scoops for applications in the food industry. VITLAB™ Polypropylene Scoops are lighter than metal, don’t dent and feature good chemical resistance. Also suitable as weighing scoops.
Brand: Vitlab™ 39394
3.96 EUR valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Additional Details : Weight : 0.04500kg
Description
- White polypropylene scoop
- With precision formed filling edge and comfortable, stable handle
- Easily readable volume quantities on the upper side of the handle
Specifications
Measuring Scoop | |
Flat | |
White | |
100 mm |
Polypropylene | |
10 mL | |
For contact with food |