Learn More
Vitlab™ Polypropylene Scoop
Product Code. 10538701
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Length (Metric):
100 mm
135 mm
160 mm
200 mm
260 mm
315 mm
385 mm
Unit Size:
Each
Pack of 12
Pack of 6
Product Code. 10538701
Brand: Vitlab™ 39494
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these corrosion resistant scoops for applications in the food industry. VITLAB™ Polypropylene Scoops are lighter than metal, don’t dent and feature good chemical resistance. Also suitable as weighing scoops.
  • White polypropylene scoop
  • With precision formed filling edge and comfortable, stable handle
  • Easily readable volume quantities on the upper side of the handle
Specifications
Capacity (Metric) 25 mL
Color White
Product Type Measuring Scoop
For Use With (Application) For contact with food
Material Polypropylene
Length (Metric) 135 mm