Learn More
Masterflex™ Polypropylene Sanitary Concentric Reducer
Click to view available options
Material:
Polypropylene
Unit Size:
Each
Description
- Meet USP Class VI for pharmaceutical use and FDA, USDA and 3A standards for food contact.
- All fittings are compatible with Tri-Clamp™; fittings.
- Excellent chemical resistance.
- Low leachable characteristics.
- concentric reducers decrease in diameter towards the center thus forming a conical cylinder.
- Though both materials may be autoclaved, PVDF tolerates high temperatures better than PP does and will perform more consistently after repeated autoclaving.
- Clamps and gaskets required for a complete system are sold separately.
Specifications
Specifications
| Description | Sanitary Connection |
| Material | Polypropylene |
| Min. Operating Temperature (English) | 15°F |
| Min. Operating Temperature (Metric) | -9°C |
| Resistance Properties | Very good chemical resistance |
| Port Size | 2 in., 1 1/2 in. |
| Sterilization Method | Autoclave or Ethylene Oxide |
| Maximum Pressure | 125 psi in ambient conditions, using plastic true union style clamp |
| Max. Operating Temperature (English) | 180°F |
| Max. Operating Temperature (Metric) | 82°F |
| Show More |