Learn More
Camlab™ Polypropylene Rack
Brand: Camlab™ RTP711096G
Additional Details : Weight : 0.08000kg
Specifications
Rack | |
Green | |
Polypropylene | |
Laboratory Sample Storage | |
1 Each | |
198 mm | |
21 mm |
Lid | |
Tubes (96) | |
Yes | |
Freezer storage system | |
-40°C to +70°C | |
129 mm |