Learn More
Camlab™ Polypropylene Rack
POA
Specifications
Product Type | Rack |
---|---|
Includes | Lid |
Material | Polypropylene |
Stackable | Yes |
For Use With (Application) | Laboratory Sample Storage |
Product Code | Brand | Color | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Holds | Price | Quantity & Availability | ||||
11757586
|
Camlab™
RTP711096N |
Natural | Tubes (96) |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11797586
|
Camlab™
RTP711096Y |
Yellow | Tubes (96) |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11840593
|
Camlab™
RTP/711048G |
Green | Tubes (48) |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11830593
|
Camlab™
RTP/711048B |
Blue | Tubes (48) |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11854653
|
Camlab™
RTP/711048N |
Natural | Tubes (48) |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11874653
|
Camlab™
RTP/711048Y |
Yellow | Tubes (48) |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11864653
|
Camlab™
RTP/711048R |
Red | Tubes (48) |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11777586
|
Camlab™
RTP711096G |
Green | Tubes (96) |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Specifications
Rack | |
Polypropylene | |
Laboratory Sample Storage | |
1 Each | |
198 mm |
Lid | |
Yes | |
Freezer storage system | |
-40°C to +70°C | |
21 mm |