Learn More
Witeg™ Polypropylene Pouring Ring
Product Code. p-8000809
Click to view available options
Color:
Blue
Green
Natural
Red
Unit Size:
Pack of 10
Product Code. 11844075
Brand: Witeg™ 0 175 202
Only null left
Add to basket
Add to basket
Use Witeg™ Polypropylene Pouring Rings to complement your laboratory bottle measurements.
  • For GL45 Thread
  • Polypropylene
  • Various Colors
  • Autoclavable
Specifications
Color Blue
Material Polypropylene
No. per Pack 10Pack
Quantity 10 Pack
Product Type Ring
Closure Material Polypropylene
Closure Type Screw Cap