Learn More
Witeg™ Polypropylene Pouring Ring
Click to view available options
Color:
Blue
Green
Natural
Red
Unit Size:
Pack of 10
Description
- For GL45 Thread
- Polypropylene
- Various Colors
- Autoclavable
Specifications
Specifications
| Color | Natural |
| Material | Polypropylene |
| No. per Pack | 10Pack |
| Quantity | 10Pack |
| Product Type | Pouring Ring |
| For Use With (Equipment) | For GL 45 thread |
| Closure Material | Polypropylene |
| Autoclavable | Autoclavable |