Learn More
Multiroir Controlec™ Polypropylene Plastic Transport Trays
Product Code. 11780594
Change view
Click to view available options
Weight (Metric):
1.6 kg
2.5 kg
3.8 kg
7 kg
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
11780594 7 kg Each
11750594 1.6 kg Each
11760594 2.5 kg Each
11770594 3.8 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11780594 Supplier Multiroir Controlec™ Supplier No. AMBD8632
Estimated Shipment: 07-12-2026
Specifications
Product Type Transport Tray
Color Blue
Material Polypropylene
Depth (Metric) 399 mm
Depth (Metric) Interior 507 mm
Height (Metric) 448 mm
Height (Metric) Interior 278 mm
Length (Metric) 809 mm
Length (Metric) Interior 686 mm
Model AMBD8632
Weight (Metric) 7 kg