Learn More
Brady™ Polypropylene and Molded Glass-filled Nylon No Hole Circuit Breaker Lockout
Product Code. 11791885 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
56 x 24.10 mm
82.60 x 40.30 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11791885 56 x 24.10 mm Each
11701895 82.60 x 40.30 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11791885 Supplier Brady™ Supplier No. 065396
  • ONLINE EXCLUSIVE PRICE
    16.70€
    14.60€ / Pack of 1
    Save 2.10€
    12% Off
Estimated Shipment: 18-11-2026
These No-Hole breaker lockouts are extremely versatile and easy to install using patented thumbscrew design - no screwdrivers needed. Simply tighten lockout securely onto switch tongue, pull cover over thumbscrew and lock cover to prevent clamp from being loosened.
  • Detachable Cleats (included) expand the range of applicable breakers.
  • Accepts lock shackles up to 7 mm in diameter.
Specifications
Product Type Circuit Breaker Lockout
Color Red
Material Glass-filled Nylon
Certifications/Compliance OSHA 1910.147
Description No Hole Circuit Breaker Lockout - 120/277 V Device
Diameter (English) Shackle 0.3 in.
Diameter (Metric) Shackle 7 mm
Dimensions (D x W x H) 25.40 x 56.01 x 24.13 mm
Dimensions (L x W) 56 x 24.10 mm
For Use With (Application) Lockout Tagout
Height (English) 0.95 in.
Height (Metric) 24.13 mm
Quantity 1 Ea.
Width (English) 2.205 in.
Width (Metric) 56.01 mm