Learn More
Brady™ Polypropylene and Molded Glass-filled Nylon No Hole Circuit Breaker Lockout
Click to view available options
Dimensions (L x W):
56 x 24.10 mm
82.60 x 40.30 mm
Unit Size:
Each
Description
- Detachable Cleats (included) expand the range of applicable breakers.
- Accepts lock shackles up to 7 mm in diameter.
Specifications
Specifications
| Product Type | Circuit Breaker Lockout |
| Color | Red |
| Material | Glass-filled Nylon |
| Certifications/Compliance | OSHA 29 CFR 1910.147 |
| Description | No Hole Circuit Breaker Lockout - 480/600 V Device |
| Diameter (English) Shackle | 0.3 in. |
| Diameter (Metric) Shackle | 7 mm |
| Dimensions (D x W x H) | 55.25 x 82.63 x 40.34 mm |
| Dimensions (L x W) | 82.60 x 40.30 mm |
| For Use With (Application) | Lockout Tagout |
| Show More |