Learn More
Fisherbrand™ Polypropylene Microtube
Product Code. 11848362
Click to view available options
Color:
Natural
Unit Size:
Pack of 1000
Product Code. 11848362
Brand: Fisherbrand™ 175510
Only null left
Add to basket
Edge
Add to basket
Use the Fisherbrand™ Polypropylene Microtube repeatedly since it is a reusable microtube made of sturdy polypropylene. The tube is ideal for general mixing and laboratory work.
  • reusable 
  • 2mL capacity
  • Nonsterile
  • Polypropylene
Specifications
Description Microtube
Autoclavable Autoclavable
Capacity (Metric) 2 mL
Color Natural
Material Polypropylene
Packaging 1000Pack
Sterility Non-sterile
Graduated No
Closure Type Flat Cap
Diameter (Metric) 10 mm