Learn More
Kartell™ Polypropylene Micropipet Tips
Click to view available options
Color:
Blue
Light Blue
Natural
Neutral
Yellow
Volume (Metric):
100 to 1000 μL
1000 to 5000 μL
2000 to 10,000 μL
5 to 200 μL
Format:
Bulk
Rack
For Use With (Equipment):
Gilson Pipetman, Nichiryo, Kartell Pipette, Socorex Pipette
Gilson Pipetman, Socorex Pipettor, Labsystem Pipettor, Thermo Scientific Finnpipette, Kartell Pipettor
Kartell Pipettor, Eppendorf, Sartorius Biohit Pipettor, Oxford, Hamilton Pipettor
Kartell Pluripet PL 1001, Eppendorf, Gilson Pipetman, Socorex Pipettor, Hamilton Pipettor
Unit Size:
Pack of 100
Pack of 250
Pack of 25000
Pack of 960
Description
- Cadmium-free polypropylene construction provides high clarity and water-repellent properties
- Specially designed for Kartell automatic pipets, but deliver excellent results with most available micropipet brands
- Autoclavable to 121°C for 20 minutes
- Precision delivery of sample, minimal sample retention with tip
- Packaging selection satisfies a variety of needs:
- bulk bag
- packed in racks
- refills for reloading racks
Specifications
Specifications
| Volume (Metric) | 2000 to 10,000 μL |
| Color | Neutral |
| Autoclavable | Autoclavable |
| Certifications/Compliance | CE, IVD |
| Format | Rack |
| Tip Style | Low Retention |
| For Use With (Equipment) | Gilson Pipetman, Socorex Pipettor, Labsystem Pipettor, Thermo Scientific Finnpipette, Kartell Pipettor |
| Material | Virgin Cadmium-free Polypropylene |