Learn More
Greiner Bio-One Polypropylene Microcentrifuge Tubes
Product Code. 11857822
Click to view available options
Includes:
N/A
Closure Color:
Natural
Unit Size:
Pack of 500
Product Code. 11857822
Brand: Greiner Bio-One 716201
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use in safe and long term storage. Greiner Bio-One Polypropylene Microcentrifuge Tubes offer a leak-free performance and minimal non-specific binding.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • chemical and temperature resistance
  • clarity coupled to a wide thermal range
  • May be centrifuged to 20,000g
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Screw cap
Closure Color Natural
Color Natural
Material Polypropylene
Includes N/A
Volume (Metric) 1.5mL