Learn More
Fisherbrand™ Crystal Micro Eppendorf™ Pipette Tips
Description
- Crystal
- For Eppendorf pipette
- 46mm length
Specifications
Specifications
| Sterility | Sterile |
| Color | Clear |
| Length (Metric) | 46 mm |
| Product Type | Pipette Tips |
| Disposable | Single-use |
| Product Line | Crystal Micro Eppendorf |
| Material | Crystal |