Learn More
Cole-Parmer™ Polypropylene Manifolds
54.30€ - 115.00€
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11775238
|
Cole-Parmer™
31522-07 |
79.7 mm |
54.30€
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
11755258
|
Cole-Parmer™
31522-25 |
186.4 mm |
115.00€
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
11725258
|
Cole-Parmer™
31522-19 |
155.9 mm |
67.90€
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
11785238
|
Cole-Parmer™
31522-09 |
106.1 mm |
62.60€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
11765258
|
Cole-Parmer™
31522-27 |
238.3 mm |
79.30€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
Specifications
Manifold | |
Polypropylene |