Learn More
Masterflex™ Polypropylene Male Pipe Adapter Barbed Fittings
Product Code. p-8006914
Click to view available options
Thread Size:
1/4 in. MNPT
1/8 in. MNPT
Unit Size:
Pack of 10
Product Code. 15277536
Brand: Masterflex™ 4062122
Only null left
Specifications
Autoclavable Autoclavable
Material Polypropylene
For Use With 1/8 x 5/32 in. Tubing I.D.
Packaging Quantity 10 Pack
Thread Size 1/8 in. MNPT
Product Type Male Pipe Adapter