Description
- Manufactured of animal-free natural polypropylene
- All parts meet tight dimensional tolerances for increased reliability and process purity
- Choose from a range of sizes, with a variety of connection options
- Good chemical resistance. Max temperature 87°C (190°F)
- Sterilize by ethylene oxide or gamma irradiation (not recommended for autoclaving)
- All female ports may be used with either slip or luer lock connections
- Plugs and caps are also available
Specifications
Specifications
| Material | Polypropylene |
| For Use With | Fits Male Luer Lock to 3/32 in. I.D. |
| For Use With (Application) | Ideal for a variety of applications, including syringes or small tubing |
| Packaging Quantity | 25 Pack |
| Product Type | Luer Adapter |