Learn More
Ark Plas Products™ Polypropylene Luer Plug
Product Code. 11712625
Click to view available options
Product Type:
Female Luer Plug
Locking Male Luer Plug
Male Slip Luer Plug
Unit Size:
Pack of 25
Product Code. 11712625
Brand: Ark Plas Products™ LPA31PP0
Only null left
Add to basket
Add to basket
Use these luers with a wide variety of medical and laboratory instruments. Ark Plas Products™ Polypropylene Luer Plugs feature conical tapers molded to ISO 594-1 and ISO 594-2 Standards for 6% luer tapers to ensure compatibility with all other luers molded to the same standard.
  • Natural polypropylene luer plugs
  • Feature barbs designed with no parting line for reliable, leak-free use with a broad variety of medical and laboratory instruments, including syringetips and needles or stopcocks and needles
  • Molded in virgin, lot-traceable material
  • Ark-Plas Luer fittings include a large selection of both male and female luers available in locking or slip tip variances, as well as locking rings, bulkhead luers for panel mounting, threaded luers, adapters and plugs
Specifications
Material Polypropylene
Min. Operating Temperature (Metric) -17°C
Packaging Quantity 25 Pack
Color Natural
Max. Operating Temperature (Metric) 135°C
Product Type Locking Male Luer Plug