Learn More
Equip Labo™ Polypropylene Laundry Gress Furniture
Click to view available options
Width (Metric):
600 mm
900 mm
150 cm
Unit Size:
Each
Specifications
Specifications
| Material | Polypropylene |
| Type | Laundry Gress Furniture |
| Width (Metric) | 900 mm |
| Depth (Metric) | 600 mm |