Learn More
VITLAB™ Polypropylene Laboratory Tray
Click to view available options
Height (Metric):
42 mm
Unit Size:
Each
Specifications
Specifications
| Color | White |
| Height (Metric) | 42 mm |
| Length (Metric) | 130 mm (Inner Bottom), 180 mm (Edge) |
| Material | Polypropylene |
| Type | Laboratory Tray |
| Width (Metric) | 180 mm (Inner Bottom), 230 mm (Edge) |
| Shape | Rectangular |