Learn More
Bohlender™ BOLA™ Polypropylene HPLC Bottle Distributor
Product Code. 10134781
Click to view available options
Diameter (Metric) Outer:
4 mm
6 mm
Unit Size:
Each
Product Code. 10134781
Brand: Bohlender™ D60808
Only null left
Add to basket
Add to basket
Use these caps with distributors for drawing or inserting liquids. Bohlender™ BOLA™ Polypropylene HPLC Bottle Distributor feature four ports with screw joints suitable for tubing up to 6.0mm O.D. Plugs for closing unused ports are included in delivery.
  • Green glass-fiber reinforced PP screw cap for bottle thread GL 45
  • Polypropylene body
  • Autoclavable
  • Very good chemical resistance
  • Available either with four ports with screw joints suitable for tubing O.D. 1.6 to 2.0mm and 2.1 to 4.0mm or with four ports with screw joints suitable for tubing O.D. 6.0 mm
Specifications
Closure Color Green
Diameter (Metric) Outer 6 mm
Material Polypropylene
Septum 1Each
Closure Material Glass Fiber
Quantity 1 Ea.