Learn More
Mutexil™ Polypropylene Hooded Combination Coveralls
Product Code. 11751585
Click to view available options
Size:
Large
X-Large
2X-Large
Color:
Blue
White
Unit Size:
Pack of 50
Product Code. 11751585
Brand: Mutexil™ 2000008L
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Disposable coveralls for maintenance, production and site visits
  • Polypropylene non-woven coverall with hood
  • Elastic wrists, ankles, and back
  • Zip fastening and two pockets
  • For maintenance, production site visits
  • Non hazardous environments
Specifications
Product Type General Purpose Coveralls
Size Large
Gender Unisex
Material Polypropylene
Color Blue
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Sterility Non-sterile
No. of Pockets 2
Sleeve Style Elastic
Thickness 40g/m2