missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Polypropylene Grid Inserts for Cryo Boxes

Product Code. 15118895
Change view
Click to view available options
Tube Array:
9 x 9
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tube Array unitSize
15118895 9 x 9 Pack of 5
10062562 9 x 9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15118895 Supplier Ratiolab™ Supplier No. 5220103

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An accessory for the safe storage and transport of deep freeze materials.

  • Autoclavable and temperature resistant from -90° to +121°C
  • Insert grids are removable for greater flexibility
  • Provided in packs of 5

Specifications

Color White
Material Polypropylene
Type Insert Grid
For Use With (Application) Storage/transport of deep freeze samples
Quantity 5/pk
Tube Array 9 x 9
Withstand Temperature (Metric) -90°C to +121°C
Dimensions (L x W x H) 133 x 133 x 30 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.