Learn More
RETSCH Polypropylene Gravity Lids
215.00€ - 367.00€
Specifications
Product Type | Gravity Lid |
---|---|
Closure Type | Gravity Lid |
Material | Polypropylene |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10084981
|
RETSCH
02.107.0323 |
For Autoclavable Plastic or Polypropylene Grinding Containers |
367.00€
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
10340632
|
RETSCH
02.107.0308 |
For Glass or Stainless Steel Grinding Containers |
367.00€
Each |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
10379601
|
RETSCH
02.107.0328 |
For GRINDOMIX GM 200 |
215.00€
Each |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
10045081
|
RETSCH
02.107.0327 |
For Grinding Containers of Autoclavable Plastic or Polypropylene |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
Gravity Lid | |
Gravity Lid | |
Polypropylene |