Learn More
Huenersdorf™ Polypropylene Graduated Beaker with Closed Handle and 2 Scales
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2000 mL
5000 mL
Unit Size:
Each
Description
- Polypropylene beaker
- Graduated
- Blue embossed scale
- Injection molded scale
- Transparent
Specifications
Specifications
| Color | Transparent |
| Diameter (Metric) Outer | 203 mm |
| Capacity (Metric) | 5000 mL |
| Material | Polypropylene |
| For Use With (Application) | For Use in The Laboratory, Workshop and Household |
| Height (Metric) | 257 mm |
| Includes | With Injection Molded Scale and Blue Embossed Scale |
| Increments | 100 mL |
| Quantity | 1 Ea. |
| Type | Beaker |
| Show More |