Learn More
Multiroir™ Polypropylene Stackable Drawer
Product Code. p-8000359
Click to view available options
Capacity (Metric):
10 L
15 L
25 L
Color:
Black
Dark Gray
Gray
White
Unit Size:
Each
Pack of 4
Pack of 6
Product Code. 30487000
Brand: Multiroir™ 18301BTSDM0BL
Store and organized items in protective polypropylene with the Multiroir™ Polypropylene Stackable Drawer. Available in white or dark gray, drawers are stackable and have a drawer-sliding capability for ease of use and spacesaving convenience.
  • Stackable
  • Raised edges for stacking and sliding ease
  • Tight fitting lid
Specifications
Capacity (Metric) 15 L
Color Dark Gray
Material Polypropylene
Shape Rectangular
Product Type Stackable Drawer
Quantity 1 Each
Style Raised Edges
Length (Metric) 370 mm
Height (Metric) 174 mm