Learn More
Polypropylene Disposal Bins
Product Code. p-8010469
Change view
Click to view available options
Capacity (Metric):
7 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
13433657 - -
13101264 7 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13433657 Supplier Daniels Healthcare Supplier No. DD672
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for cytotoxic and cytostatic waste which, to meet regulations, should be disposed of separately from other healthcare waste. The SHARPSGUARD™ cyto lid assists safe, easy collection of items including giving sets, syringes and gloves. Color-coded with a purple lid, handle and label, it can be safely transported for incineration after closure. Absorbent pads are included. The container is made to BS EN ISO 23907.