Learn More
Polypropylene Disposal Bins
Product Code. 13101264
Click to view available options
Capacity (Metric):
7 L
Unit Size:
Each
Product Code. 13101264
Brand: Daniels Healthcare DD673
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ideal for cytotoxic and cytostatic waste which, to meet regulations, should be disposed of separately from other healthcare waste. The SHARPSGUARD™ cyto lid assists safe, easy collection of items including giving sets, syringes and gloves. Color-coded with a purple lid, handle and label, it can be safely transported for incineration after closure. Absorbent pads are included. The container is made to BS EN ISO 23907.
Specifications
Capacity (Metric) 7 L