Learn More
Kartell™ Polypropylene Disposable Tubes
Click to view available options
Capacity (Metric):
3 mL
Unit Size:
Pack of 5000
Description
- Polypropylene
- CE and IVD certification
- Max. RCF: 3500xg
Specifications
Specifications
| Certifications/Compliance | CE IVD |
| Capacity (Metric) | 3 mL |
| Disposable | Single-use |
| Material | Polypropylene |
| Diameter (Metric) | 11.5 mm |
| Product Type | Centrifuge Tube |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur