Learn More
Fisherbrand™ Polypropylene Disposable Standard Pipette Tips
Product Code. 10677541
Click to view available options
Volume (Metric):
200 μL
1000 μL
Color:
Blue
Yellow
Length (Metric):
4.8cm
5cm
7cm
7.3cm
Unit Size:
Pack of 1000
Product Code. 10677541
Brand: Fisherbrand™ 161004
Only null left
Add to basket
Add to basket
Use FIsherbrand™ Polypropylene Disposable Standard Pipette Tips in the laboratory. Made of first-class polypropylene, they are disposable, autoclavable and packaged in a sterile bag.
  • Autoclavable
  • Disposable
  • Polypropylene
  • Packaged in a sterile bag
Specifications
Volume (Metric) 200 μL
Color Yellow
Autoclavable Autoclavable
Length (Metric) 4.8cm
Tip Style Universal
Disposable Single-use
For Use With (Equipment) Eppendorf, Sartorius Biohit