Learn More
Fisherbrand™ Polypropylene Disposable Standard Pipette Tips
Description
- Autoclavable
- Disposable
- Polypropylene
- Packaged in a sterile bag
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Color | Blue |
| Autoclavable | Autoclavable |
| Length (Metric) | 7.3cm |
| Tip Style | Universal |
| Disposable | Single-use |
| For Use With (Equipment) | Gilson Pipetman, Eppendorf, Thermo Scientific Finnpipette, Sartorius Biohit Pipettor, Handrop, Nichiryo, Socorex Pipettor |
| Material | Polypropylene |