Specifications
| Description | Conical bottom |
| Quantity | 1/Ea. |
| Size | NS 29/32 |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Product Type | Ground Joint Stopper |
| Shape | Square |