Learn More
Macherey-Nagel™ Polypropylene Chromabond Column
Click to view available options
Column Capacity:
6 mL
70 mL
Unit Size:
Pack of 30
Specifications
Specifications
| Quantity | 30 Pack |
| Type | Column |
| Column Capacity | 70 mL |
| Material | Polypropylene |