Learn More
Macherey-Nagel™ Polypropylene Chromabond™ C8 SPE Column
Product Code. 10115091
Click to view available options
Bed Weight:
100 mg
200 mg
500 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
Unit Size:
Each
Pack of 100
Pack of 50
Product Code. 10115091
Brand: Macherey-Nagel™ 730021
Specifications
Base Material Silica
Carbon Load 8%
Column Capacity (Metric) 1 mL
Column Type SPE
Bed Weight 100 mg
For Use With (Application) Pesticides, PCBs Applications
Particle Size 45 μm
Type Solid Phase Extraction Column
Endcapped No
Pore Size 60 μm
Surface Area (Metric) 500 m2/g