Learn More
Ratiolab™ Polypropylene Blue Pipet Tip E
Product Code. 10789521
Click to view available options
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 120
Pack of 960
Product Code. 10789521
Brand: Ratiolab™ 2100610
Use these pipet tips for simple and safe liquid handling in the most sensitive applications. Ratiolab™ Polypropylene Blue Pipet Tips E are ideal for manual or electronic, single or multichannel pipets.
  • Ideal for all commercial pipets
  • Made of high quality  polypropylene
  • Tight fit due to the precisely fitting cone
  • Optimal handling due to stable shape
  • Autoclavable
  • Guaranteed dust free due to special ionization treatment
  • Available in pre-sterilized or non-sterile
  • Excellentr4 transparency and high quality surfaces
  • Minimal residual wetting
  • Excellent volume accuracy due to precise tip orifice
  • Practice-oriented tip volumes
Specifications
Volume (Metric) 100 to 1000 μL
Color Blue
Autoclavable Autoclavable
Length (Metric) 6.9cm
Certifications/Compliance CE, IVD
For Use With (Application) Ideal for All Commercially Available Pippets
Sterilization Method Autoclavable
Format Bulk
For Use With (Equipment) Universal Fit