Learn More
Semadeni™ Color-Coded Plastic Beakers
Product Code. p-8000779
Click to view available options
Color:
Blue
Green
Natural
Yellow
Unit Size:
Pack of 100
Product Code. 11793835
Brand: Semadeni™ 8609
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Excellent for the storage, mixing or pouring of liquids in the laboratory. Semadeni™ Color-Coded Plastic Beakers are manufactured from durable polypropylene.
  • Translucent colors
  • 30mL capacity
  • Graduated
  • Optional LDPE lids available for purchase
Specifications
Color Natural
Capacity (Metric) 30 mL
Material Polypropylene
For Use With (Application) Suitable as Dosing or Laboratory Beakers
Quantity 100 Pk.
Type Beaker
Disposable Single-use
Graduated Yes