Learn More
Masterflex™ Polypropylene Barbed Fitting
Click to view available options
For Use With:
2.4mm
Unit Size:
Pack of 10
Specifications
Specifications
| Diameter (Metric) Inner | 2.4 mm |
| Material | Polypropylene |
| For Use With | 2.4mm |
| Min. Operating Temperature (Metric) | 0°C |
| Packaging Quantity | 10 Pack |
| Port Size | 19 x 7.93mm |
| Max. Operating Temperature (Metric) | 135°C |
| Product Type | Barbed Fitting |